Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
Chromosome Location
Chr.6: 39335100 - 39725405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289020.1 NP_001275949.1 8 - 9 1076 78
NM_001289021.1 NP_001275950.1 8 - 9 1076 78
NM_145027.4 NP_659464.3 8 - 9 1076 78
XM_005248904.4 XP_005248961.1 8 - 9 1076 78
XM_011514357.2 XP_011512659.1 8 - 9 1001 78
XM_011514358.2 XP_011512660.1 8 - 9 1001 78
XM_011514359.2 XP_011512661.1 8 - 9 1001 78
XM_011514361.2 XP_011512663.1 8 - 9 1076 78
XM_017010427.1 XP_016865916.1 7 - 8 966 78
XM_017010428.1 XP_016865917.1 3 - 4 426 78
GenBank mRNA AK131471.1 - 8 - 9 1076 78
BC117307.1 - 8 - 9 1006 78
BC143444.1 - 8 - 9 1006 78
BC143445.1 - 8 - 9 1006 78
BX649045.1 - 5 - 6 656 78

View Full Product Details