Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINC00574
Entrez Gene ID
80069
Gene Name
long intergenic non-protein coding RNA 574
Gene Aliases
C6orf208, dJ182D15.1
Chromosome Location
Chr.6: 169790073 - 169802873 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026780.1 1 - 2 339 119
GenBank mRNA AK023224.1 - 1 - 2 91 119
DA918406.1 - 1 - 2 339 119


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AU132009.1 - 1 - 2 91 Chr. 6 - 169790321 - 169801290 [+] on Build GRCh38 119
GenBankmRNA DB259408.1 - 1 - 2 346 Chr. 6 - 169790066 - 169800892 [+] on Build GRCh38 119
GenBankmRNA BJ991487.1 - 1 - 2 96 Chr. 6 - 169790316 - 169801132 [+] on Build GRCh38 119
GenBankmRNA DA533671.1 - 1 - 2 90 Chr. 6 - 169790326 - 169801098 [+] on Build GRCh38 119


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 242 LINC00242 C6orf122, NCRNA00242, dJ266L20.5 Anti-sense Chr. - 169788790 - 169798825 on Build GRCh38
Downstream uncharacterized LOC100131532 LOC100131532 167.3kb Sense Chr. - 169962623 - 169967678 on Build GRCh38

View Full Product Details