Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARPBP
Entrez Gene ID
55010
Gene Name
PARP1 binding protein
Gene Aliases
AROM, C12orf48, PARI
Chromosome Location
Chr.12: 102120171 - 102197520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319988.1 NP_001306917.1 5 - 6 851 159
NM_001319993.1 NP_001306922.1 5 - 6 728 159
NM_001319994.1 NP_001306923.1 4 - 5 615 159
NM_001319995.1 NP_001306924.1 5 - 6 773 159
NM_017915.4 NP_060385.3 4 - 5 620 159
XM_011538509.1 XP_011536811.1 3 - 4 386 159
XM_011538514.2 XP_011536816.1 3 - 4 433 159
XM_017019535.1 XP_016875024.1 3 - 4 459 159
XM_017019536.1 XP_016875025.1 4 - 5 620 159
XM_017019537.1 XP_016875026.1 4 - 5 620 159
XM_017019538.1 XP_016875027.1 4 - 5 620 159
XM_017019539.1 XP_016875028.1 3 - 4 386 159
XM_017019540.1 XP_016875029.1 4 - 5 582 159
XM_017019541.1 XP_016875030.1 4 - 5 620 159
XM_017019542.1 XP_016875031.1 4 - 5 620 159
XM_017019543.1 XP_016875032.1 4 - 5 614 159
XM_017019544.1 XP_016875033.1 4 - 5 614 159
GenBank mRNA AK000648.1 - 4 - 5 539 159
AK302724.1 - 5 - 6 837 159
AK303571.1 - 4 - 5 546 159
BC030962.1 - 5 - 6 672 159
BC070270.1 - 4 - 5 576 159
BC099734.1 - 3 - 4 402 159
BC099844.1 - 3 - 4 402 159

View Full Product Details