The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002463.1 | NP_002454.1 | 9 - 10 | 1371 | 115 |
XM_005260983.4 | XP_005261040.1 | 9 - 10 | 1469 | 115 | |
XM_005260984.1 | XP_005261041.1 | 9 - 10 | 1649 | 115 | |
XM_011529571.1 | XP_011527873.1 | 9 - 10 | 1404 | 115 | |
XM_011529572.1 | XP_011527874.1 | 9 - 10 | 1507 | 115 | |
XM_011529573.1 | XP_011527875.1 | 8 - 9 | 1273 | 115 | |
XM_011529574.2 | XP_011527876.1 | 4 - 5 | 551 | 115 | |
GenBank mRNA | AK122952.1 | - | 4 - 5 | 1557 | 115 |
BC035293.1 | - | 8 - 9 | 1333 | 115 | |
M30818.1 | - | 9 - 10 | 1371 | 115 | |
M33883.1 | - | 8 - 9 | 1184 | 115 |