Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PTPRD
- Entrez Gene ID
-
5789
- Gene Name
- protein tyrosine phosphatase, receptor type D
- Gene Aliases
-
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
- Chromosome Location
-
Chr.9: 8314246 - 10612840 on Build GRCh38
View Full Product Details