Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRD
Entrez Gene ID
5789
Gene Name
protein tyrosine phosphatase, receptor type D
Gene Aliases
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location
Chr.9: 8314246 - 10612840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040712.2 NP_001035802.1 10 - 11 1770 79
NM_001171025.1 NP_001164496.1 9 - 10 1749 79
NM_002839.3 NP_002830.1 22 - 23 2419 79
NM_130391.3 NP_569075.2 11 - 12 1779 79
NM_130392.3 NP_569076.2 11 - 12 1779 79
NM_130393.3 NP_569077.2 10 - 11 1761 79
XM_006716817.3 XP_006716880.1 21 - 22 2717 79
XM_006716823.2 XP_006716886.1 21 - 22 2711 79
XM_006716825.3 XP_006716888.1 21 - 22 2713 79
XM_006716827.3 XP_006716890.1 18 - 19 2671 79
XM_006716832.3 XP_006716895.1 19 - 20 2675 79
XM_006716833.3 XP_006716896.1 21 - 22 2707 79
XM_006716834.3 XP_006716897.1 18 - 19 2669 79
XM_006716835.3 XP_006716898.1 20 - 21 2698 79
XM_006716837.3 XP_006716900.1 20 - 21 2687 79
XM_006716838.3 XP_006716901.1 19 - 20 2680 79
XM_006716839.3 XP_006716902.1 18 - 19 2666 79
XM_011517992.2 XP_011516294.1 21 - 22 2710 79
XM_017014958.1 XP_016870447.1 21 - 22 2716 79
XM_017014959.1 XP_016870448.1 22 - 23 2788 79
XM_017014960.1 XP_016870449.1 20 - 21 2677 79
XM_017014961.1 XP_016870450.1 20 - 21 2661 79
XM_017014962.1 XP_016870451.1 12 - 13 1849 79
XM_017014963.1 XP_016870452.1 21 - 22 2716 79
XM_017014964.1 XP_016870453.1 21 - 22 2715 79
XM_017014965.1 XP_016870454.1 21 - 22 2718 79
XM_017014966.1 XP_016870455.1 20 - 21 2707 79
XM_017014967.1 XP_016870456.1 20 - 21 2702 79
XM_017014968.1 XP_016870457.1 21 - 22 2713 79
XM_017014969.1 XP_016870458.1 20 - 21 2698 79
XM_017014970.1 XP_016870459.1 21 - 22 2716 79
XM_017014971.1 XP_016870460.1 19 - 20 2687 79
XM_017014972.1 XP_016870461.1 21 - 22 2716 79
XM_017014973.1 XP_016870462.1 19 - 20 2683 79
XM_017014974.1 XP_016870463.1 21 - 22 2716 79
XM_017014975.1 XP_016870464.1 18 - 19 2672 79
XM_017014976.1 XP_016870465.1 21 - 22 2713 79
XM_017014977.1 XP_016870466.1 21 - 22 2714 79
XM_017014978.1 XP_016870467.1 21 - 22 2713 79
XM_017014979.1 XP_016870468.1 21 - 22 2715 79
XM_017014980.1 XP_016870469.1 21 - 22 2710 79
XM_017014981.1 XP_016870470.1 18 - 19 2666 79
XM_017014982.1 XP_016870471.1 21 - 22 2712 79
XM_017014983.1 XP_016870472.1 21 - 22 2712 79
XM_017014984.1 XP_016870473.1 20 - 21 2703 79
XM_017014985.1 XP_016870474.1 21 - 22 2709 79
XM_017014986.1 XP_016870475.1 21 - 22 2708 79
XM_017014987.1 XP_016870476.1 20 - 21 2690 79
XM_017014988.1 XP_016870477.1 20 - 21 2690 79
XM_017014989.1 XP_016870478.1 21 - 22 2708 79
XM_017014990.1 XP_016870479.1 21 - 22 2707 79
XM_017014991.1 XP_016870480.1 21 - 22 2707 79
XM_017014992.1 XP_016870481.1 20 - 21 2698 79
XM_017014993.1 XP_016870482.1 21 - 22 2705 79
XM_017014994.1 XP_016870483.1 21 - 22 2705 79
XM_017014995.1 XP_016870484.1 20 - 21 2702 79
GenBank mRNA AB208869.1 - 9 - 10 1596 79
AB211400.1 - 21 - 22 2387 79
BC106713.1 - 9 - 10 1697 79
BC106714.1 - 11 - 12 1727 79
BC106715.2 - 11 - 12 1727 79
BC106716.1 - 10 - 11 1709 79
L38929.1 - 12 - 13 1829 79
X54133.1 - 2 - 3 509 79

View Full Product Details