The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242498.1 | NP_001229427.1 | 6 - 7 | 699 | 56 |
NM_001242500.1 | NP_001229429.1 | 5 - 6 | 601 | 56 | |
NM_001242501.1 | NP_001229430.1 | 4 - 5 | 499 | 56 | |
NM_020679.3 | NP_065730.2 | 6 - 7 | 678 | 56 | |
XM_005257530.1 | XP_005257587.1 | 6 - 7 | 820 | 56 | |
XM_005257532.4 | XP_005257589.1 | 5 - 6 | 576 | 56 | |
XM_005257533.1 | XP_005257590.1 | 5 - 6 | 675 | 56 | |
XM_011525051.2 | XP_011523353.1 | 6 - 7 | 915 | 56 | |
XM_011525052.2 | XP_011523354.1 | 6 - 7 | 894 | 56 | |
XM_011525053.2 | XP_011523355.1 | 5 - 6 | 792 | 56 | |
XM_011525054.2 | XP_011523356.1 | 5 - 6 | 759 | 56 | |
XM_011525055.2 | XP_011523357.1 | 4 - 5 | 636 | 56 | |
XM_011525057.2 | XP_011523359.1 | 3 - 4 | 339 | 56 | |
XM_017024893.1 | XP_016880382.1 | 5 - 6 | 1071 | 56 | |
XM_017024894.1 | XP_016880383.1 | 5 - 6 | 706 | 56 | |
XM_017024895.1 | XP_016880384.1 | 5 - 6 | 522 | 56 | |
GenBank mRNA | AK300711.1 | - | 6 - 7 | 638 | 56 |
BC033759.1 | - | 5 - 6 | 577 | 56 | |
BU190758.1 | - | 5 - 6 | 600 | 56 | |
CA488406.1 | - | 4 - 5 | 499 | 56 | |
KU178731.1 | - | 5 - 6 | 541 | 56 | |
KU178732.1 | - | 5 - 6 | 562 | 56 | |
KU178733.1 | - | 4 - 5 | 439 | 56 | |
KU178734.1 | - | 3 - 4 | 283 | 56 |