The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002518.3 | NP_002509.2 | 12 - 13 | 1418 | 88 |
XM_005263953.2 | XP_005264010.1 | 12 - 13 | 1329 | 88 | |
XM_005263959.2 | XP_005264016.1 | 12 - 13 | 1329 | 88 | |
XM_005263960.2 | XP_005264017.1 | 9 - 10 | 1020 | 88 | |
XM_005263961.4 | XP_005264018.1 | 10 - 11 | 1223 | 88 | |
XM_011511242.2 | XP_011509544.1 | 11 - 12 | 1296 | 88 | |
XM_011511243.2 | XP_011509545.1 | 12 - 13 | 1329 | 88 | |
XM_017004214.1 | XP_016859703.1 | 12 - 13 | 1329 | 88 | |
XM_017004215.1 | XP_016859704.1 | 12 - 13 | 1329 | 88 | |
XM_017004216.1 | XP_016859705.1 | 12 - 13 | 1329 | 88 | |
XM_017004217.1 | XP_016859706.1 | 9 - 10 | 1020 | 88 | |
GenBank mRNA | AK226093.1 | - | 7 - 8 | 746 | 88 |
AK294733.1 | - | 12 - 13 | 1329 | 88 | |
BC028107.1 | - | 11 - 12 | 1161 | 88 | |
BC051351.1 | - | 12 - 13 | 1382 | 88 | |
BC072383.1 | - | 12 - 13 | 1385 | 88 | |
BM724652.1 | - | 5 - 6 | 513 | 88 | |
BX640699.1 | - | 1 - 2 | 466 | 88 | |
KJ534892.1 | - | 11 - 12 | 1132 | 88 | |
U51625.1 | - | 11 - 12 | 1167 | 88 | |
U77970.1 | - | 12 - 13 | 1418 | 88 |