The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002518.3 | NP_002509.2 | 6 - 7 | 775 | 85 |
XM_005263953.2 | XP_005264010.1 | 6 - 7 | 686 | 85 | |
XM_005263959.2 | XP_005264016.1 | 6 - 7 | 686 | 85 | |
XM_005263960.2 | XP_005264017.1 | 6 - 7 | 686 | 85 | |
XM_005263961.4 | XP_005264018.1 | 4 - 5 | 580 | 85 | |
XM_011511242.2 | XP_011509544.1 | 5 - 6 | 653 | 85 | |
XM_011511243.2 | XP_011509545.1 | 6 - 7 | 686 | 85 | |
XM_017004214.1 | XP_016859703.1 | 6 - 7 | 686 | 85 | |
XM_017004215.1 | XP_016859704.1 | 6 - 7 | 686 | 85 | |
XM_017004216.1 | XP_016859705.1 | 6 - 7 | 686 | 85 | |
XM_017004217.1 | XP_016859706.1 | 6 - 7 | 686 | 85 | |
GenBank mRNA | AK226093.1 | - | 1 - 2 | 103 | 85 |
AK294733.1 | - | 6 - 7 | 686 | 85 | |
BC028107.1 | - | 5 - 6 | 518 | 85 | |
BC051351.1 | - | 6 - 7 | 739 | 85 | |
BC072383.1 | - | 6 - 7 | 742 | 85 | |
KJ534892.1 | - | 5 - 6 | 489 | 85 | |
U77970.1 | - | 6 - 7 | 775 | 85 |