Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUL4A
Entrez Gene ID
8451
Gene Name
cullin 4A
Gene Aliases
-
Chromosome Location
Chr.13: 113208193 - 113265078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008895.2 NP_001008895.1 10 - 11 1163 98
NM_001278513.1 NP_001265442.1 10 - 11 1307 98
NM_001278514.1 NP_001265443.1 10 - 11 975 98
NM_003589.2 NP_003580.1 10 - 11 989 98
XM_011537523.2 XP_011535825.1 9 - 10 1523 98
GenBank mRNA AB012193.1 - 4 - 5 337 98
AB178950.1 - 10 - 11 1040 98
AF077188.1 - 10 - 11 900 98
AK296700.1 - 2 - 3 284 98
AK314835.1 - 10 - 11 1262 98
AL833355.1 - 9 - 10 927 98
AY365124.1 - 10 - 11 1040 98
BC008308.1 - 10 - 11 1051 98

View Full Product Details