The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008895.2 | NP_001008895.1 | 16 - 17 | 1872 | 88 |
NM_001278513.1 | NP_001265442.1 | 16 - 17 | 2016 | 88 | |
NM_001278514.1 | NP_001265443.1 | 16 - 17 | 1684 | 88 | |
NM_003589.2 | NP_003580.1 | 16 - 17 | 1698 | 88 | |
XM_011537523.2 | XP_011535825.1 | 15 - 16 | 2232 | 88 | |
GenBank mRNA | AB012193.1 | - | 10 - 11 | 1046 | 88 |
AF077188.1 | - | 16 - 17 | 1609 | 88 | |
AK296700.1 | - | 8 - 9 | 993 | 88 | |
AK314835.1 | - | 16 - 17 | 1971 | 88 | |
AL833355.1 | - | 15 - 16 | 1636 | 88 | |
AY365124.1 | - | 16 - 17 | 1749 | 88 | |
BC008308.1 | - | 16 - 17 | 1760 | 88 | |
U58090.1 | - | 6 - 7 | 736 | 88 |