The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199775.1 | NP_001186704.1 | 9 - 10 | 1646 | 137 |
NM_001304.4 | NP_001295.2 | 9 - 10 | 2284 | 137 | |
GenBank mRNA | AK302497.1 | - | 9 - 10 | 1888 | 137 |
AK302878.1 | - | 9 - 10 | 1605 | 137 | |
AK316409.1 | - | 9 - 10 | 1646 | 137 | |
BC045549.1 | - | 9 - 10 | 2270 | 137 | |
BC045624.1 | - | 9 - 10 | 2258 | 137 | |
BC051702.1 | - | 9 - 10 | 2284 | 137 | |
D85390.1 | - | 9 - 10 | 2262 | 137 | |
U65090.3 | - | 9 - 10 | 2242 | 137 |