Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
Chromosome Location
Chr.21: 25880550 - 26171128 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000484.3 NP_000475.1 6 - 7 1061 111
NM_001136016.3 NP_001129488.1 6 - 7 1042 111
NM_001136130.2 NP_001129602.1 5 - 6 893 111
NM_001204301.1 NP_001191230.1 6 - 7 1061 111
NM_001204302.1 NP_001191231.1 6 - 7 1061 111
NM_201413.2 NP_958816.1 6 - 7 1061 111
GenBank mRNA AK294534.1 - 5 - 6 840 111
AK295621.1 - 6 - 7 1003 111
AK312326.1 - 6 - 7 926 111
BC065529.1 - 6 - 7 992 111
M28373.1 - 4 - 5 624 111
X06989.1 - 6 - 7 985 111

View Full Product Details