The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320412.1 | NP_001307341.1 | 2 - 3 | 287 | 130 |
NM_006657.2 | NP_006648.1 | 2 - 3 | 287 | 130 | |
NM_206965.1 | NP_996848.1 | 2 - 3 | 287 | 130 | |
XM_006723961.3 | XP_006724024.2 | 2 - 3 | 578 | 130 | |
XM_006723962.3 | XP_006724025.2 | 2 - 3 | 1186 | 130 | |
XM_011529434.2 | XP_011527736.1 | 2 - 3 | 1201 | 130 | |
XM_011529435.2 | XP_011527737.1 | 2 - 3 | 585 | 130 | |
XM_011529436.2 | XP_011527738.1 | 2 - 3 | 1186 | 130 | |
XM_011529437.2 | XP_011527739.1 | 2 - 3 | 580 | 130 | |
XM_011529440.2 | XP_011527742.1 | 2 - 3 | 572 | 130 | |
GenBank mRNA | AF169017.1 | - | 2 - 3 | 287 | 130 |
AF289021.1 | - | 2 - 3 | 287 | 130 | |
AF289022.1 | - | 2 - 3 | 287 | 130 | |
AF289023.1 | - | 2 - 3 | 287 | 130 | |
AF289024.1 | - | 2 - 3 | 287 | 130 | |
BC052248.1 | - | 2 - 3 | 283 | 130 | |
BC136383.1 | - | 2 - 3 | 279 | 130 | |
BC136395.1 | - | 2 - 3 | 279 | 130 |