Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTCN1
Entrez Gene ID
79679
Gene Name
V-set domain containing T cell activation inhibitor 1
Gene Aliases
B7-H4, B7H4, B7S1, B7X, B7h.5, PRO1291, VCTN1
Chromosome Location
Chr.1: 117143587 - 117210992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253849.1 NP_001240778.1 3 - 4 631 118
NM_024626.3 NP_078902.2 3 - 4 549 118
NR_045603.1 3 - 4 639 118
XM_011542143.2 XP_011540445.2 3 - 4 833 118
XM_017002335.1 XP_016857824.1 4 - 5 859 118
GenBank mRNA AK026071.1 - 3 - 4 514 118
AK225413.1 - 3 - 4 516 118
AK303466.1 - 3 - 4 631 118
AY280972.1 - 3 - 4 444 118
AY346100.1 - 3 - 4 504 118
AY358352.1 - 3 - 4 504 118
BC065717.1 - 3 - 4 610 118
BC074729.2 - 3 - 4 475 118
BX648021.1 - 4 - 5 533 118
CN259811.1 - 1 - 2 117 118
DB198367.1 - 3 - 4 516 118
DQ836392.1 - 3 - 4 537 118

View Full Product Details