The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253849.1 | NP_001240778.1 | 4 - 5 | 910 | 124 |
NM_001253850.1 | NP_001240779.1 | 3 - 4 | 480 | 124 | |
NM_024626.3 | NP_078902.2 | 4 - 5 | 828 | 124 | |
NR_045603.1 | 4 - 5 | 918 | 124 | ||
NR_045604.1 | 4 - 5 | 622 | 124 | ||
XM_011542143.2 | XP_011540445.2 | 4 - 5 | 1112 | 124 | |
XM_017002335.1 | XP_016857824.1 | 5 - 6 | 1138 | 124 | |
GenBank mRNA | AK026071.1 | - | 4 - 5 | 793 | 124 |
AK225413.1 | - | 4 - 5 | 795 | 124 | |
AK303466.1 | - | 4 - 5 | 910 | 124 | |
AK310812.1 | - | 4 - 5 | 589 | 124 | |
AY280972.1 | - | 4 - 5 | 723 | 124 | |
AY346100.1 | - | 4 - 5 | 783 | 124 | |
AY358352.1 | - | 4 - 5 | 783 | 124 | |
BC065717.1 | - | 4 - 5 | 889 | 124 | |
BC074729.2 | - | 4 - 5 | 754 | 124 | |
BX648021.1 | - | 5 - 6 | 812 | 124 | |
CN259811.1 | - | 2 - 3 | 396 | 124 | |
DQ103757.1 | - | 3 - 4 | 375 | 124 |