The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164115.1 | NP_001157587.1 | 2 - 3 | 662 | 100 |
NM_001164116.1 | NP_001157588.1 | 3 - 4 | 686 | 100 | |
NM_020356.3 | NP_065089.2 | 2 - 3 | 662 | 100 | |
GenBank mRNA | AJ276678.1 | - | 3 - 4 | 686 | 100 |
AK027760.1 | - | 2 - 3 | 659 | 100 | |
AK308057.1 | - | 2 - 3 | 663 | 100 | |
AK314653.1 | - | 2 - 3 | 595 | 100 | |
BC027951.1 | - | 2 - 3 | 612 | 100 |