The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308157.1 | NP_001295086.1 | 6 - 7 | 1313 | 64 |
NM_016524.3 | NP_057608.2 | 6 - 7 | 1504 | 64 | |
XM_005255362.3 | XP_005255419.1 | 6 - 7 | 1467 | 64 | |
XM_011545865.1 | XP_011544167.1 | 6 - 7 | 1140 | 64 | |
XM_011545866.2 | XP_011544168.1 | 6 - 7 | 2043 | 64 | |
XM_011545868.1 | XP_011544170.1 | 5 - 6 | 1137 | 64 | |
XM_011545869.2 | XP_011544171.1 | 6 - 7 | 1486 | 64 | |
XM_011545870.2 | XP_011544172.1 | 6 - 7 | 1366 | 64 | |
XM_011545871.2 | XP_011544173.1 | 6 - 7 | 1472 | 64 | |
XM_011545872.2 | XP_011544174.1 | 5 - 6 | 1449 | 64 | |
XM_017023280.1 | XP_016878769.1 | 6 - 7 | 1498 | 64 | |
XM_017023281.1 | XP_016878770.1 | 5 - 6 | 1172 | 64 | |
GenBank mRNA | AF220560.1 | - | 6 - 7 | 1078 | 64 |
AK098747.1 | - | 6 - 7 | 1313 | 64 | |
AK226136.1 | - | 4 - 5 | 4677 | 64 | |
AK296000.1 | - | 4 - 5 | 1193 | 64 | |
BC004518.2 | - | 6 - 7 | 1080 | 64 |