The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001382.3 | NP_001373.2 | 1 - 2 | 587 | 99 |
XM_005271422.2 | XP_005271479.1 | 1 - 2 | 603 | 99 | |
XM_017017294.1 | XP_016872783.1 | 1 - 2 | 603 | 99 | |
GenBank mRNA | AK128572.1 | - | 3 - 4 | 1719 | 99 |
AK225994.1 | - | 1 - 2 | 332 | 99 | |
AK312783.1 | - | 1 - 2 | 390 | 99 | |
BC000325.2 | - | 1 - 2 | 335 | 99 | |
BT006802.1 | - | 1 - 2 | 167 | 99 | |
Z82022.1 | - | 1 - 2 | 246 | 99 |