The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005355.3 | NP_005346.3 | 3 - 4 | 430 | 63 |
NM_030615.2 | NP_085118.2 | 3 - 4 | 430 | 63 | |
XM_011535802.2 | XP_011534104.1 | 5 - 6 | 537 | 63 | |
XM_011535803.2 | XP_011534105.1 | 5 - 6 | 537 | 63 | |
GenBank mRNA | AB012722.2 | - | 3 - 4 | 430 | 63 |
AK289488.1 | - | 5 - 6 | 683 | 63 | |
BC130489.1 | - | 3 - 4 | 478 | 63 |