The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197941.3 | NP_922932.2 | 19 - 20 | 3290 | 109 |
NR_135689.1 | 20 - 21 | 3441 | 109 | ||
XM_011543113.2 | XP_011541415.1 | 19 - 20 | 3305 | 109 | |
XM_011543114.2 | XP_011541416.1 | 19 - 20 | 2884 | 109 | |
XM_011543115.2 | XP_011541417.1 | 18 - 19 | 3193 | 109 | |
XM_011543116.2 | XP_011541418.1 | 17 - 18 | 3052 | 109 | |
XM_011543117.1 | XP_011541419.1 | 17 - 18 | 2643 | 109 | |
XM_011543119.2 | XP_011541421.1 | 19 - 20 | 3334 | 109 | |
XM_011543120.2 | XP_011541422.1 | 19 - 20 | 3331 | 109 | |
XM_011543121.2 | XP_011541423.2 | 15 - 16 | 1671 | 109 | |
XM_011543123.2 | XP_011541425.1 | 13 - 14 | 1409 | 109 | |
XM_011543124.2 | XP_011541426.1 | 10 - 11 | 1193 | 109 | |
XM_011543125.1 | XP_011541427.1 | 9 - 10 | 1048 | 109 | |
GenBank mRNA | AB209685.1 | - | 20 - 21 | 2938 | 109 |
AF140674.1 | - | 17 - 18 | 2310 | 109 | |
AY692424.1 | - | 19 - 20 | 3248 | 109 | |
AY692425.1 | - | 19 - 20 | 2589 | 109 | |
AY692426.1 | - | 18 - 19 | 2456 | 109 |