The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042555.2 | NP_001036020.1 | 8 - 9 | 919 | 94 |
NM_001278351.1 | NP_001265280.1 | 9 - 10 | 1040 | 94 | |
NM_001278353.1 | NP_001265282.1 | 9 - 10 | 983 | 94 | |
NM_001278354.1 | NP_001265283.1 | 8 - 9 | 972 | 94 | |
NM_001278355.1 | NP_001265284.1 | 8 - 9 | 954 | 94 | |
NM_001278356.1 | NP_001265285.1 | 7 - 8 | 858 | 94 | |
NM_001278357.1 | NP_001265286.1 | 5 - 6 | 719 | 94 | |
NM_006654.4 | NP_006645.3 | 8 - 9 | 926 | 94 | |
XM_017018717.1 | XP_016874206.1 | 9 - 10 | 1006 | 94 | |
XM_017018718.1 | XP_016874207.1 | 9 - 10 | 1000 | 94 | |
XM_017018719.1 | XP_016874208.1 | 4 - 5 | 465 | 94 | |
GenBank mRNA | AF036717.1 | - | 3 - 4 | 421 | 94 |
AK312477.1 | - | 7 - 8 | 777 | 94 | |
BC021562.1 | - | 5 - 6 | 662 | 94 | |
DA762876.1 | - | 1 - 2 | 48 | 94 |