Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC17
Entrez Gene ID
23390
Gene Name
zinc finger DHHC-type containing 17
Gene Aliases
HIP14, HIP3, HSPC294, HYPH
Chromosome Location
Chr.12: 76764074 - 76853701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015336.2 NP_056151.2 2 - 3 354 73
XM_005268750.4 XP_005268807.1 2 - 3 194 73
XM_011538083.1 XP_011536385.1 2 - 3 218 73
XM_017019091.1 XP_016874580.1 2 - 3 194 73
XM_017019092.1 XP_016874581.1 1 - 2 98 73
GenBank mRNA AB023163.1 - 2 - 3 298 73
AB024494.1 - 2 - 3 841 73
AB097013.1 - 2 - 3 174 73
AB097035.1 - 2 - 3 174 73
AF049612.1 - 2 - 3 187 73
AF161412.1 - 2 - 3 186 73
AK292866.1 - 2 - 3 170 73
BC050324.1 - 2 - 3 313 73
DA227374.1 - 2 - 3 354 73

View Full Product Details