Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC17
Entrez Gene ID
23390
Gene Name
zinc finger DHHC-type containing 17
Gene Aliases
HIP14, HIP3, HSPC294, HYPH
Chromosome Location
Chr.12: 76764074 - 76853701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015336.2 NP_056151.2 7 - 8 935 105
XM_005268750.4 XP_005268807.1 7 - 8 734 105
XM_011538083.1 XP_011536385.1 7 - 8 799 105
XM_017019094.1 XP_016874583.1 6 - 7 615 105
XM_017019095.1 XP_016874584.1 3 - 4 327 105
GenBank mRNA AB023163.1 - 7 - 8 879 105
AB024494.1 - 7 - 8 1425 105
AB097013.1 - 7 - 8 755 105
AB097035.1 - 7 - 8 755 105
AK292866.1 - 7 - 8 751 105
BC050324.1 - 7 - 8 894 105

View Full Product Details