The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015336.2 | NP_056151.2 | 8 - 9 | 1059 | 114 |
XM_005268750.4 | XP_005268807.1 | 8 - 9 | 858 | 114 | |
XM_011538083.1 | XP_011536385.1 | 8 - 9 | 923 | 114 | |
XM_017019091.1 | XP_016874580.1 | 9 - 10 | 1009 | 114 | |
XM_017019092.1 | XP_016874581.1 | 8 - 9 | 872 | 114 | |
XM_017019093.1 | XP_016874582.1 | 8 - 9 | 821 | 114 | |
XM_017019094.1 | XP_016874583.1 | 7 - 8 | 739 | 114 | |
XM_017019095.1 | XP_016874584.1 | 4 - 5 | 451 | 114 | |
GenBank mRNA | AB023163.1 | - | 8 - 9 | 1003 | 114 |
AB024494.1 | - | 8 - 9 | 1549 | 114 | |
AB097013.1 | - | 8 - 9 | 879 | 114 | |
AB097035.1 | - | 8 - 9 | 879 | 114 | |
AK292866.1 | - | 8 - 9 | 875 | 114 | |
AK299089.1 | - | 8 - 9 | 881 | 114 | |
BC050324.1 | - | 8 - 9 | 1018 | 114 |