Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCGBP
Entrez Gene ID
8857
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
FC(GAMMA)BP
Chromosome Location
Chr.19: 39863323 - 39934626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003890.2 NP_003881.2 - 15942 84
GenBank mRNA BU731683.1 - 2 - 3 84
BX113070.1 - 2 - 3 519 84
D84239.1 - - 15942 84

View Full Product Details