Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
Chromosome Location
Chr.7: 151085831 - 151144436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042535.3 NP_001036000.1 2 - 3 708 98
NM_001308304.1 NP_001295233.1 2 - 3 708 98
NM_031946.6 NP_114152.3 2 - 3 708 98
XM_017011734.1 XP_016867223.1 2 - 3 147 98
GenBank mRNA AB209781.1 - 2 - 3 149 98
AF359283.3 - 3 - 4 283 98
AF413079.1 - 2 - 3 283 98
BC020426.1 - 1 - 2 64 98
BC069236.1 - 3 - 4 280 98

View Full Product Details