The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251845.1 | NP_001238774.1 | 8 - 9 | 1569 | 141 |
NM_003304.4 | NP_003295.1 | 7 - 8 | 1467 | 141 | |
XM_005247738.3 | XP_005247795.1 | 7 - 8 | 1244 | 141 | |
XM_005247739.2 | XP_005247796.1 | 6 - 7 | 1195 | 141 | |
XM_017007121.1 | XP_016862610.1 | 8 - 9 | 2177 | 141 | |
XM_017007122.1 | XP_016862611.1 | 6 - 7 | 2092 | 141 | |
GenBank mRNA | AB255425.1 | - | 7 - 8 | 1331 | 141 |
BC112338.1 | - | 8 - 9 | 1386 | 141 | |
BC113953.1 | - | 8 - 9 | 1384 | 141 | |
X89066.1 | - | 7 - 8 | 1468 | 141 |