The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001115113.2 | NP_001108585.2 | 8 - 9 | 1445 | 90 |
NM_025160.6 | NP_079436.4 | 8 - 9 | 1493 | 90 | |
XM_005273273.4 | XP_005273330.1 | 8 - 9 | 1394 | 90 | |
XM_006711817.3 | XP_006711880.1 | 8 - 9 | 1391 | 90 | |
XM_011544281.2 | XP_011542583.1 | 8 - 9 | 1234 | 90 | |
XM_011544282.2 | XP_011542584.1 | 5 - 6 | 556 | 90 | |
XM_017002396.1 | XP_016857885.1 | 8 - 9 | 1082 | 90 | |
GenBank mRNA | AB209887.1 | - | 8 - 9 | 1493 | 90 |
AK024669.1 | - | 4 - 5 | 507 | 90 | |
AK225061.1 | - | 4 - 5 | 507 | 90 | |
AK299230.1 | - | 5 - 6 | 678 | 90 | |
AY221751.1 | - | 8 - 9 | 1283 | 90 | |
AY304473.1 | - | 8 - 9 | 1050 | 90 | |
BC031471.2 | - | 3 - 4 | 436 | 90 | |
BC052301.1 | - | 7 - 8 | 889 | 90 | |
BC063817.1 | - | 4 - 5 | 555 | 90 | |
EF011612.1 | - | 7 - 8 | 888 | 90 |