The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001115113.2 | NP_001108585.2 | 12 - 13 | 1916 | 151 |
NM_025160.6 | NP_079436.4 | 12 - 13 | 1964 | 151 | |
XM_006711817.3 | XP_006711880.1 | 12 - 13 | 1862 | 151 | |
XM_011544281.2 | XP_011542583.1 | 12 - 13 | 1705 | 151 | |
XM_011544282.2 | XP_011542584.1 | 9 - 10 | 1027 | 151 | |
XM_017002396.1 | XP_016857885.1 | 12 - 13 | 1553 | 151 | |
GenBank mRNA | AB209887.1 | - | 12 - 13 | 1964 | 151 |
AK024669.1 | - | 8 - 9 | 978 | 151 | |
AK225061.1 | - | 8 - 9 | 978 | 151 | |
AK299230.1 | - | 9 - 10 | 1149 | 151 | |
AY221751.1 | - | 12 - 13 | 1754 | 151 | |
AY304473.1 | - | 12 - 13 | 1521 | 151 | |
BC031471.2 | - | 7 - 8 | 907 | 151 | |
BC034498.1 | - | 14 - 15 | 1956 | 151 | |
BC052301.1 | - | 11 - 12 | 1360 | 151 | |
BC063817.1 | - | 8 - 9 | 1026 | 151 | |
EF011612.1 | - | 11 - 12 | 1359 | 151 |