Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNH7
Entrez Gene ID
90134
Gene Name
potassium voltage-gated channel subfamily H member 7
Gene Aliases
ERG3, HERG3, Kv11.3
Chromosome Location
Chr.2: 162371407 - 162838747 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033272.3 NP_150375.2 15 - 16 3547 114
XM_011512109.2 XP_011510411.1 16 - 17 3461 114
XM_017005218.1 XP_016860707.1 16 - 17 3452 114
XM_017005219.1 XP_016860708.1 15 - 16 3428 114
XM_017005220.1 XP_016860709.1 14 - 15 3416 114
GenBank mRNA AF032897.1 - 15 - 16 3418 114

View Full Product Details