Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DENND3
Entrez Gene ID
22898
Gene Name
DENN domain containing 3
Gene Aliases
-
Chromosome Location
Chr.8: 141128618 - 141195804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014957.2 NP_055772.2 19 - 20 3285 186
XM_005250838.4 XP_005250895.2 19 - 20 3176 186
XM_005250839.2 XP_005250896.2 19 - 20 3176 186
XM_005250840.4 XP_005250897.2 18 - 19 3020 186
XM_005250841.3 XP_005250898.2 18 - 19 3020 186
XM_005250842.4 XP_005250899.1 17 - 18 3037 186
XM_011516933.1 XP_011515235.1 19 - 20 3176 186
XM_011516934.2 XP_011515236.1 19 - 20 3176 186
XM_011516938.2 XP_011515240.1 12 - 13 2331 186
XM_011516939.2 XP_011515241.1 10 - 11 1854 186
XM_011516940.2 XP_011515242.1 10 - 11 1831 186
XM_017013241.1 XP_016868730.1 19 - 20 3178 186
XM_017013242.1 XP_016868731.1 20 - 21 3539 186
XM_017013243.1 XP_016868732.1 11 - 12 2291 186
GenBank mRNA AB020677.2 - 14 - 15 2472 186
AK091556.1 - 1 - 2 800 186
AK131505.1 - 2 - 3 954 186
BC133023.1 - 17 - 18 2872 186
BC144226.1 - 17 - 18 2872 186

View Full Product Details