Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAT2A
Entrez Gene ID
4144
Gene Name
methionine adenosyltransferase 2A
Gene Aliases
MATA2, MATII, SAMS2
Chromosome Location
Chr.2: 85538978 - 85545280 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005911.5 NP_005902.1 5 - 6 857 64
GenBank mRNA AK291126.1 - 5 - 6 665 64
AK293841.1 - 5 - 6 667 64
AK297758.1 - 5 - 6 560 64
AK308758.1 - 5 - 6 525 64
AK309338.1 - 5 - 6 665 64
AK316411.1 - 5 - 6 815 64
BC001686.2 - 5 - 6 654 64
BC001854.2 - 5 - 6 654 64
DQ083239.1 - 5 - 6 593 64
X68836.1 - 5 - 6 612 64

View Full Product Details