Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1K
Entrez Gene ID
152926
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1K
Gene Aliases
BDP, MSUDMV, PP2Ckappa, PP2Cm, PTMP, UG0882E07
Chromosome Location
Chr.4: 88257609 - 88284831 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152542.4 NP_689755.3 6 - 7 1470 114
XM_006714111.3 XP_006714174.1 6 - 7 1531 114
XM_017007803.1 XP_016863292.1 6 - 7 1118 114
XM_017007804.1 XP_016863293.1 7 - 8 1658 114
XM_017007805.1 XP_016863294.1 8 - 9 1658 114
XM_017007806.1 XP_016863295.1 6 - 7 991 114
XM_017007807.1 XP_016863296.1 5 - 6 709 114
GenBank mRNA AK054678.1 - 6 - 7 1198 114
AK314417.1 - 6 - 7 1195 114
AL834271.1 - 6 - 7 1189 114
AX766363.1 - 5 - 6 985 114
AY157615.1 - 6 - 7 1198 114
AY435431.1 - 6 - 7 1375 114
AY994097.1 - 6 - 7 1206 114
BC037552.1 - 6 - 7 1234 114

View Full Product Details