The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301248.1 | NP_001288177.1 | 14 - 15 | 2942 | 112 |
NM_178039.3 | NP_829883.1 | 14 - 15 | 2973 | 112 | |
NM_178040.3 | NP_829884.1 | 15 - 16 | 3057 | 112 | |
NR_027946.2 | 14 - 15 | 3018 | 112 | ||
NR_027948.2 | 15 - 16 | 3150 | 112 | ||
NR_027949.2 | 11 - 12 | 2169 | 112 | ||
XM_011520941.2 | XP_011519243.1 | 15 - 16 | 3101 | 112 | |
XM_017019052.1 | XP_016874541.1 | 17 - 18 | 3242 | 112 | |
XM_017019053.1 | XP_016874542.1 | 17 - 18 | 3176 | 112 | |
XM_017019054.1 | XP_016874543.1 | 17 - 18 | 3102 | 112 | |
XM_017019055.1 | XP_016874544.1 | 17 - 18 | 3098 | 112 | |
XM_017019056.1 | XP_016874545.1 | 17 - 18 | 3039 | 112 | |
XM_017019057.1 | XP_016874546.1 | 17 - 18 | 3233 | 112 | |
XM_017019058.1 | XP_016874547.1 | 17 - 18 | 3167 | 112 | |
XM_017019059.1 | XP_016874548.1 | 16 - 17 | 3230 | 112 | |
XM_017019060.1 | XP_016874549.1 | 16 - 17 | 3221 | 112 | |
XM_017019061.1 | XP_016874550.1 | 16 - 17 | 3188 | 112 | |
XM_017019062.1 | XP_016874551.1 | 16 - 17 | 3179 | 112 | |
XM_017019063.1 | XP_016874552.1 | 15 - 16 | 3176 | 112 | |
XM_017019064.1 | XP_016874553.1 | 15 - 16 | 3167 | 112 | |
XM_017019065.1 | XP_016874554.1 | 16 - 17 | 3149 | 112 | |
XM_017019066.1 | XP_016874555.1 | 15 - 16 | 3095 | 112 | |
XM_017019067.1 | XP_016874556.1 | 14 - 15 | 3083 | 112 | |
XM_017019068.1 | XP_016874557.1 | 15 - 16 | 3167 | 112 | |
XM_017019069.1 | XP_016874558.1 | 15 - 16 | 3056 | 112 | |
XM_017019070.1 | XP_016874559.1 | 14 - 15 | 3035 | 112 | |
XM_017019071.1 | XP_016874560.1 | 14 - 15 | 2963 | 112 | |
XM_017019072.1 | XP_016874561.1 | 17 - 18 | 3242 | 112 | |
XM_017019074.1 | XP_016874563.1 | 10 - 11 | 1222 | 112 | |
XM_017019075.1 | XP_016874564.1 | 9 - 10 | 1166 | 112 | |
GenBank mRNA | AB015617.1 | - | 14 - 15 | 2976 | 112 |
AB029004.2 | - | 14 - 15 | 2837 | 112 | |
AB053468.1 | - | 15 - 16 | 3091 | 112 | |
AB053469.1 | - | 11 - 12 | 2104 | 112 | |
AB053470.1 | - | 14 - 15 | 2915 | 112 | |
AB053471.1 | - | 15 - 16 | 2967 | 112 | |
AK000148.1 | - | 2 - 3 | 307 | 112 | |
AK092201.1 | - | 4 - 5 | 592 | 112 | |
AK294351.1 | - | 12 - 13 | 1937 | 112 | |
BC037377.1 | - | 4 - 5 | 580 | 112 | |
BC132782.1 | - | 14 - 15 | 2918 | 112 | |
BC132784.1 | - | 14 - 15 | 2918 | 112 | |
BC144287.1 | - | 14 - 15 | 2918 | 112 | |
BC150248.1 | - | 14 - 15 | 2858 | 112 |