Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ERC1
- Entrez Gene ID
-
23085
- Gene Name
- ELKS/RAB6-interacting/CAST family member 1
- Gene Aliases
-
Cast2, ELKS, ERC-1, RAB6IP2
- Chromosome Location
-
Chr.12: 990010 - 1495933 on Build GRCh38
View Full Product Details