Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERC1
Entrez Gene ID
23085
Gene Name
ELKS/RAB6-interacting/CAST family member 1
Gene Aliases
Cast2, ELKS, ERC-1, RAB6IP2
Chromosome Location
Chr.12: 990010 - 1495933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301248.1 NP_001288177.1 7 - 8 1895 67
NM_178039.3 NP_829883.1 7 - 8 1926 67
NM_178040.3 NP_829884.1 8 - 9 2010 67
NR_027946.2 7 - 8 2091 67
NR_027948.2 7 - 8 2091 67
NR_027949.2 3 - 4 1110 67
XM_011520941.2 XP_011519243.1 8 - 9 2054 67
XM_017019052.1 XP_016874541.1 8 - 9 2129 67
XM_017019053.1 XP_016874542.1 8 - 9 2063 67
XM_017019054.1 XP_016874543.1 8 - 9 1989 67
XM_017019055.1 XP_016874544.1 8 - 9 1985 67
XM_017019056.1 XP_016874545.1 8 - 9 1926 67
XM_017019057.1 XP_016874546.1 8 - 9 2120 67
XM_017019058.1 XP_016874547.1 8 - 9 2054 67
XM_017019059.1 XP_016874548.1 8 - 9 2129 67
XM_017019060.1 XP_016874549.1 8 - 9 2120 67
XM_017019061.1 XP_016874550.1 8 - 9 2129 67
XM_017019062.1 XP_016874551.1 8 - 9 2120 67
XM_017019063.1 XP_016874552.1 8 - 9 2129 67
XM_017019064.1 XP_016874553.1 8 - 9 2120 67
XM_017019065.1 XP_016874554.1 7 - 8 2036 67
XM_017019066.1 XP_016874555.1 7 - 8 2036 67
XM_017019067.1 XP_016874556.1 7 - 8 2036 67
XM_017019068.1 XP_016874557.1 8 - 9 2120 67
XM_017019069.1 XP_016874558.1 8 - 9 2129 67
XM_017019070.1 XP_016874559.1 8 - 9 2120 67
XM_017019071.1 XP_016874560.1 7 - 8 2036 67
XM_017019072.1 XP_016874561.1 8 - 9 2129 67
XM_017019073.1 XP_016874562.1 8 - 9 2129 67
GenBank mRNA AB015617.1 - 7 - 8 2049 67
AB029004.2 - 6 - 7 1778 67
AB053468.1 - 7 - 8 2032 67
AB053469.1 - 3 - 4 1045 67
AB053470.1 - 7 - 8 1868 67
AB053471.1 - 8 - 9 1920 67
AK091577.1 - 7 - 8 2036 67
BC068006.1 - 6 - 7 1777 67
BC132782.1 - 7 - 8 1871 67
BC132784.1 - 7 - 8 1871 67
BC144287.1 - 7 - 8 1871 67
BC150248.1 - 6 - 7 1799 67

View Full Product Details