The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258008.1 | NP_001244937.1 | 4 - 5 | 655 | 115 |
NM_017984.4 | NP_060454.3 | 4 - 5 | 655 | 115 | |
XM_005250479.2 | XP_005250536.1 | 4 - 5 | 1086 | 115 | |
XM_005250480.2 | XP_005250537.1 | 4 - 5 | 1086 | 115 | |
XM_006716035.3 | XP_006716098.1 | 4 - 5 | 1086 | 115 | |
XM_006716036.2 | XP_006716099.1 | 4 - 5 | 590 | 115 | |
XM_006716037.3 | XP_006716100.1 | 4 - 5 | 1086 | 115 | |
XM_006716038.3 | XP_006716101.1 | 4 - 5 | 1086 | 115 | |
XM_006716040.3 | XP_006716103.1 | 4 - 5 | 1086 | 115 | |
XM_017012379.1 | XP_016867868.1 | 4 - 5 | 1086 | 115 | |
XM_017012380.1 | XP_016867869.1 | 4 - 5 | 590 | 115 | |
GenBank mRNA | AK093038.1 | - | 4 - 5 | 590 | 115 |
AK300745.1 | - | 4 - 5 | 511 | 115 | |
AK316553.1 | - | 4 - 5 | 843 | 115 | |
AL136735.1 | - | 4 - 5 | 526 | 115 | |
BC002725.1 | - | 1 - 2 | 244 | 115 |