The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032656.3 | NP_116045.2 | 21 - 22 | 2965 | 126 |
XM_005253590.3 | XP_005253647.1 | 21 - 22 | 2969 | 126 | |
GenBank mRNA | AB040950.1 | - | 18 - 19 | 2337 | 126 |
AK123968.1 | - | 1 - 2 | 1035 | 126 | |
BC002575.2 | - | 18 - 19 | 2348 | 126 | |
BC037964.1 | - | 21 - 22 | 2947 | 126 |