The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032656.3 | NP_116045.2 | 24 - 25 | 3318 | 89 |
XM_005253590.3 | XP_005253647.1 | 24 - 25 | 3322 | 89 | |
GenBank mRNA | AB040950.1 | - | 21 - 22 | 2690 | 89 |
AK123968.1 | - | 3 - 4 | 1527 | 89 | |
BC002575.2 | - | 21 - 22 | 2701 | 89 | |
BC037964.1 | - | 24 - 25 | 3300 | 89 |