Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHX37
Entrez Gene ID
57647
Gene Name
DEAH-box helicase 37
Gene Aliases
DDX37
Chromosome Location
Chr.12: 124946824 - 124989125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032656.3 NP_116045.2 25 - 26 3391 67
XM_005253590.3 XP_005253647.1 25 - 26 3395 67
GenBank mRNA AB040950.1 - 22 - 23 2763 67
AK123968.1 - 4 - 5 1600 67
BC002575.2 - 22 - 23 2774 67
BC037964.1 - 25 - 26 3373 67

View Full Product Details