Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT81
Entrez Gene ID
28981
Gene Name
intraflagellar transport 81
Gene Aliases
CDV-1, CDV-1R, CDV1, CDV1R, DV1
Chromosome Location
Chr.12: 110124335 - 110218795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014055.3 NP_054774.2 1 - 2 526 117
NM_031473.3 NP_113661.2 1 - 2 526 117
XM_017019218.1 XP_016874707.1 1 - 2 526 117
GenBank mRNA AF332010.1 - 1 - 2 484 117
AK291043.1 - 1 - 2 428 117
BC004536.2 - 1 - 2 362 117

View Full Product Details