The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143779.1 | NP_001137251.1 | 9 - 10 | 1068 | 117 |
NM_014055.3 | NP_054774.2 | 9 - 10 | 1486 | 117 | |
NM_031473.3 | NP_113661.2 | 9 - 10 | 1486 | 117 | |
XM_011538232.2 | XP_011536534.1 | 9 - 10 | 1014 | 117 | |
XM_017019217.1 | XP_016874706.1 | 9 - 10 | 1178 | 117 | |
GenBank mRNA | AF139540.1 | - | 9 - 10 | 1026 | 117 |
AF250326.1 | - | 8 - 9 | 979 | 117 | |
AF332010.1 | - | 9 - 10 | 1444 | 117 | |
AK000874.1 | - | 2 - 3 | 248 | 117 | |
AK091549.1 | - | 8 - 9 | 846 | 117 | |
AK291043.1 | - | 9 - 10 | 1388 | 117 | |
BC004536.2 | - | 9 - 10 | 1322 | 117 | |
BC108257.1 | - | 8 - 9 | 951 | 117 |