Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
Chromosome Location
Chr.22: 19966491 - 20016802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001670.2 NP_001661.1 10 - 11 2161 58
XM_006724243.2 XP_006724306.1 10 - 11 2177 58
XM_006724245.2 XP_006724308.1 10 - 11 2177 58
XM_006724246.3 XP_006724309.1 8 - 9 5013 58
XM_006724247.3 XP_006724310.1 8 - 9 1821 58
XM_006724249.3 XP_006724312.1 10 - 11 2177 58
XM_011530180.1 XP_011528482.1 10 - 11 2177 58
XM_011530182.2 XP_011528484.1 5 - 6 789 58
GenBank mRNA BC137436.1 - 9 - 10 1939 58
U51269.1 - 10 - 11 2162 58

View Full Product Details