Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
Chromosome Location
Chr.22: 19966491 - 20016802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001670.2 NP_001661.1 16 - 17 2909 119
XM_005261242.2 XP_005261299.1 15 - 16 2907 119
XM_005261243.3 XP_005261300.1 15 - 16 2907 119
XM_005261244.3 XP_005261301.1 15 - 16 2907 119
XM_006724243.2 XP_006724306.1 16 - 17 2925 119
XM_006724245.2 XP_006724308.1 16 - 17 2925 119
XM_006724246.3 XP_006724309.1 14 - 15 5761 119
XM_006724247.3 XP_006724310.1 14 - 15 2569 119
XM_006724248.3 XP_006724311.1 13 - 14 2550 119
XM_011530179.2 XP_011528481.1 15 - 16 2892 119
XM_011530180.1 XP_011528482.1 16 - 17 2925 119
XM_011530182.2 XP_011528484.1 11 - 12 1537 119
XM_011530183.2 XP_011528485.1 9 - 10 1889 119
GenBank mRNA AK092664.1 - 9 - 10 1356 119
BC137436.1 - 15 - 16 2687 119
U51269.1 - 16 - 17 2910 119

View Full Product Details