Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARVCF
Entrez Gene ID
421
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases
-
Chromosome Location
Chr.22: 19966491 - 20016802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001670.2 NP_001661.1 17 - 18 2969 133
XM_005261243.3 XP_005261300.1 16 - 17 2967 133
XM_006724247.3 XP_006724310.1 15 - 16 2629 133
XM_006724248.3 XP_006724311.1 14 - 15 2610 133
XM_011530180.1 XP_011528482.1 17 - 18 2985 133
GenBank mRNA BC137436.1 - 16 - 17 2747 133
U51269.1 - 17 - 18 2970 133

View Full Product Details