Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4G3
Entrez Gene ID
8672
Gene Name
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location
Chr.1: 20806292 - 21177320 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198801.1 NP_001185730.1 13 - 14 2444 105
NM_001198802.1 NP_001185731.1 14 - 15 2465 105
NM_003760.4 NP_003751.2 11 - 12 2081 105
XM_011542356.2 XP_011540658.1 16 - 17 3044 105
XM_011542361.2 XP_011540663.1 16 - 17 3044 105
XM_011542370.1 XP_011540672.1 16 - 17 3047 105
XM_011542371.1 XP_011540673.1 14 - 15 2789 105
XM_011542374.1 XP_011540676.1 13 - 14 2756 105
XM_011542375.2 XP_011540677.1 13 - 14 4344 105
XM_011542381.2 XP_011540683.1 3 - 4 430 105
XM_017002672.1 XP_016858161.1 17 - 18 4582 105
XM_017002673.1 XP_016858162.1 15 - 16 3026 105
XM_017002674.1 XP_016858163.1 15 - 16 3023 105
XM_017002675.1 XP_016858164.1 15 - 16 3014 105
XM_017002676.1 XP_016858165.1 14 - 15 2993 105
XM_017002677.1 XP_016858166.1 14 - 15 2870 105
XM_017002678.1 XP_016858167.1 16 - 17 3047 105
XM_017002679.1 XP_016858168.1 15 - 16 2933 105
XM_017002680.1 XP_016858169.1 12 - 13 1963 105
XM_017002681.1 XP_016858170.1 15 - 16 2930 105
XM_017002682.1 XP_016858171.1 15 - 16 3023 105
XM_017002683.1 XP_016858172.1 14 - 15 2912 105
XM_017002684.1 XP_016858173.1 15 - 16 4438 105
XM_017002685.1 XP_016858174.1 15 - 16 3014 105
XM_017002686.1 XP_016858175.1 15 - 16 3011 105
XM_017002687.1 XP_016858176.1 14 - 15 2900 105
XM_017002688.1 XP_016858177.1 11 - 12 1929 105
XM_017002689.1 XP_016858178.1 16 - 17 3047 105
XM_017002690.1 XP_016858179.1 14 - 15 2897 105
XM_017002691.1 XP_016858180.1 14 - 15 2993 105
XM_017002692.1 XP_016858181.1 13 - 14 2879 105
XM_017002693.1 XP_016858182.1 12 - 13 2750 105
XM_017002694.1 XP_016858183.1 15 - 16 2933 105
XM_017002695.1 XP_016858184.1 15 - 16 2930 105
XM_017002696.1 XP_016858185.1 14 - 15 2912 105
XM_017002697.1 XP_016858186.1 14 - 15 2909 105
XM_017002698.1 XP_016858187.1 14 - 15 2900 105
XM_017002699.1 XP_016858188.1 16 - 17 3047 105
XM_017002700.1 XP_016858189.1 13 - 14 2879 105
XM_017002701.1 XP_016858190.1 13 - 14 2876 105
XM_017002702.1 XP_016858191.1 12 - 13 2699 105
XM_017002703.1 XP_016858192.1 11 - 12 1971 105
XM_017002704.1 XP_016858193.1 15 - 16 3023 105
XM_017002705.1 XP_016858194.1 15 - 16 2207 105
XM_017002706.1 XP_016858195.1 15 - 16 2207 105
XM_017002707.1 XP_016858196.1 12 - 13 2039 105
XM_017002708.1 XP_016858197.1 12 - 13 2039 105
XM_017002709.1 XP_016858198.1 13 - 14 2791 105
XM_017002710.1 XP_016858199.1 16 - 17 3047 105
XM_017002711.1 XP_016858200.1 16 - 17 3047 105
GenBank mRNA AB209119.1 - 14 - 15 2424 105
AF012072.2 - 11 - 12 2081 105
AK294883.1 - 5 - 6 1328 105
AK302873.1 - 1 - 2 211 105
BC094683.1 - 9 - 10 1088 105
BC136643.1 - 11 - 12 2004 105

View Full Product Details