Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4G3
Entrez Gene ID
8672
Gene Name
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location
Chr.1: 20806292 - 21177320 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198801.1 NP_001185730.1 24 - 25 3999 83
NM_001198802.1 NP_001185731.1 24 - 25 3909 83
NM_003760.4 NP_003751.2 21 - 22 3525 83
XM_011542356.2 XP_011540658.1 27 - 28 4599 83
XM_011542361.2 XP_011540663.1 26 - 27 4488 83
XM_011542370.1 XP_011540672.1 27 - 28 4602 83
XM_011542371.1 XP_011540673.1 25 - 26 4344 83
XM_011542374.1 XP_011540676.1 24 - 25 4311 83
XM_011542375.2 XP_011540677.1 24 - 25 5899 83
XM_011542381.2 XP_011540683.1 14 - 15 1985 83
XM_017002672.1 XP_016858161.1 28 - 29 6137 83
XM_017002673.1 XP_016858162.1 26 - 27 4581 83
XM_017002674.1 XP_016858163.1 26 - 27 4578 83
XM_017002675.1 XP_016858164.1 26 - 27 4569 83
XM_017002676.1 XP_016858165.1 25 - 26 4548 83
XM_017002677.1 XP_016858166.1 25 - 26 4425 83
XM_017002678.1 XP_016858167.1 26 - 27 4491 83
XM_017002679.1 XP_016858168.1 26 - 27 4488 83
XM_017002680.1 XP_016858169.1 23 - 24 3518 83
XM_017002681.1 XP_016858170.1 26 - 27 4485 83
XM_017002682.1 XP_016858171.1 25 - 26 4467 83
XM_017002683.1 XP_016858172.1 25 - 26 4467 83
XM_017002684.1 XP_016858173.1 26 - 27 5993 83
XM_017002685.1 XP_016858174.1 25 - 26 4458 83
XM_017002686.1 XP_016858175.1 25 - 26 4455 83
XM_017002687.1 XP_016858176.1 25 - 26 4455 83
XM_017002688.1 XP_016858177.1 22 - 23 3484 83
XM_017002689.1 XP_016858178.1 27 - 28 4602 83
XM_017002690.1 XP_016858179.1 25 - 26 4452 83
XM_017002691.1 XP_016858180.1 24 - 25 4437 83
XM_017002692.1 XP_016858181.1 24 - 25 4434 83
XM_017002693.1 XP_016858182.1 23 - 24 4305 83
XM_017002694.1 XP_016858183.1 25 - 26 4377 83
XM_017002695.1 XP_016858184.1 25 - 26 4374 83
XM_017002696.1 XP_016858185.1 24 - 25 4356 83
XM_017002697.1 XP_016858186.1 24 - 25 4353 83
XM_017002698.1 XP_016858187.1 24 - 25 4344 83
XM_017002699.1 XP_016858188.1 26 - 27 4491 83
XM_017002700.1 XP_016858189.1 23 - 24 4323 83
XM_017002701.1 XP_016858190.1 23 - 24 4320 83
XM_017002702.1 XP_016858191.1 23 - 24 4254 83
XM_017002703.1 XP_016858192.1 21 - 22 3415 83
XM_017002704.1 XP_016858193.1 25 - 26 4467 83
XM_017002705.1 XP_016858194.1 26 - 27 3762 83
XM_017002706.1 XP_016858195.1 25 - 26 3651 83
XM_017002707.1 XP_016858196.1 23 - 24 3594 83
XM_017002708.1 XP_016858197.1 22 - 23 3483 83
XM_017002709.1 XP_016858198.1 24 - 25 4346 83
GenBank mRNA AB209119.1 - 24 - 25 3868 83
AF012072.2 - 21 - 22 3525 83
AK294883.1 - 15 - 16 2772 83
AK302873.1 - 12 - 13 1766 83
BC094683.1 - 19 - 20 2532 83
BC136643.1 - 21 - 22 3448 83
Z34918.1 - 5 - 6 611 83

View Full Product Details