Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4G3
Entrez Gene ID
8672
Gene Name
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location
Chr.1: 20806292 - 21177320 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198801.1 NP_001185730.1 4 - 5 556 71
NM_001198802.1 NP_001185731.1 4 - 5 556 71
NM_001198803.2 NP_001185732.1 4 - 5 345 71
NM_003760.4 NP_003751.2 2 - 3 190 71
XM_011542356.2 XP_011540658.1 4 - 5 988 71
XM_011542361.2 XP_011540663.1 4 - 5 988 71
XM_011542370.1 XP_011540672.1 4 - 5 988 71
XM_011542371.1 XP_011540673.1 4 - 5 865 71
XM_011542374.1 XP_011540676.1 4 - 5 865 71
XM_011542375.2 XP_011540677.1 4 - 5 2453 71
XM_017002673.1 XP_016858162.1 4 - 5 988 71
XM_017002674.1 XP_016858163.1 4 - 5 988 71
XM_017002675.1 XP_016858164.1 4 - 5 988 71
XM_017002676.1 XP_016858165.1 4 - 5 988 71
XM_017002677.1 XP_016858166.1 4 - 5 865 71
XM_017002678.1 XP_016858167.1 4 - 5 988 71
XM_017002679.1 XP_016858168.1 4 - 5 988 71
XM_017002681.1 XP_016858170.1 4 - 5 988 71
XM_017002682.1 XP_016858171.1 4 - 5 988 71
XM_017002683.1 XP_016858172.1 4 - 5 988 71
XM_017002684.1 XP_016858173.1 5 - 6 2514 71
XM_017002685.1 XP_016858174.1 4 - 5 988 71
XM_017002686.1 XP_016858175.1 4 - 5 988 71
XM_017002687.1 XP_016858176.1 4 - 5 988 71
XM_017002689.1 XP_016858178.1 4 - 5 988 71
XM_017002690.1 XP_016858179.1 4 - 5 988 71
XM_017002691.1 XP_016858180.1 4 - 5 988 71
XM_017002692.1 XP_016858181.1 4 - 5 988 71
XM_017002694.1 XP_016858183.1 4 - 5 988 71
XM_017002695.1 XP_016858184.1 4 - 5 988 71
XM_017002696.1 XP_016858185.1 4 - 5 988 71
XM_017002697.1 XP_016858186.1 4 - 5 988 71
XM_017002698.1 XP_016858187.1 4 - 5 988 71
XM_017002699.1 XP_016858188.1 4 - 5 988 71
XM_017002700.1 XP_016858189.1 4 - 5 988 71
XM_017002701.1 XP_016858190.1 4 - 5 988 71
XM_017002702.1 XP_016858191.1 4 - 5 988 71
XM_017002704.1 XP_016858193.1 4 - 5 988 71
XM_017002705.1 XP_016858194.1 4 - 5 988 71
XM_017002706.1 XP_016858195.1 4 - 5 988 71
XM_017002707.1 XP_016858196.1 4 - 5 988 71
XM_017002708.1 XP_016858197.1 4 - 5 988 71
XM_017002709.1 XP_016858198.1 4 - 5 988 71
XM_017002710.1 XP_016858199.1 4 - 5 988 71
XM_017002711.1 XP_016858200.1 4 - 5 988 71
XM_017002712.1 XP_016858201.1 5 - 6 2514 71
GenBank mRNA AB209119.1 - 4 - 5 515 71
AF012072.2 - 2 - 3 190 71
AK225489.1 - 1 - 2 124 71
AK302087.1 - 2 - 3 200 71
BC030578.1 - 6 - 7 438 71
BC094683.1 - 1 - 2 37 71
BC136643.1 - 1 - 2 95 71
DB042721.1 - 5 - 6 398 71
DB096818.1 - 4 - 5 315 71

View Full Product Details