Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FARS2
Entrez Gene ID
10667
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD14, FARS1, HSPC320, PheRS
Chromosome Location
Chr.6: 5261001 - 5771592 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318872.1 NP_001305801.1 6 - 7 1580 83
NM_006567.4 NP_006558.1 6 - 7 1574 83
XM_005248812.3 XP_005248869.1 7 - 8 1638 83
XM_017010186.1 XP_016865675.1 6 - 7 2357 83
XM_017010187.1 XP_016865676.1 7 - 8 2359 83
GenBank mRNA AF097441.1 - 5 - 6 1230 83
AI338405.1 - 1 - 2 83
AK223423.1 - 6 - 7 1374 83
AK312454.1 - 6 - 7 1554 83
BC020239.1 - 6 - 7 1374 83
BC021112.1 - 6 - 7 1374 83
BG212473.1 - 1 - 2 158 83
CA432122.1 - 1 - 2 83
CR542279.1 - 5 - 6 1218 83

View Full Product Details